Skip to Content
Merck
trans-1,2-Cyclohexanedicarboxylic anhydride 97%

trans-1,2-Cyclohexanedicarboxylic anhydride

trans-1,2-Cyclohexanedicarboxylic anhydride 97%
Synonym(s):
trans-Cyclohexane-1,2-dicarboxylic acid anhydride, trans-Hexahydro-1,3-isobenzofurandione, trans-Hexahydrophthalic anhydride
Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing