
Thulium(III) trifluoromethanesulfonate

Synonym(s):
Thulium(III) triflate, Trifluoromethanesulfonic acid thulium(III) salt
Linear Formula:
(CF3SO3)3Tm
CAS Number:
Molecular Weight:
616.14
Products
Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing