Skip to Content
Merck
Thio-L-citrulline ≥98.0%

Thio-L-citrulline

Thio-L-citrulline ≥98.0%
Synonym(s):
(S)-2-Amino-5-(thioureido)pentanoic acid, N5-Aminothioxomethyl-L-ornithine
Empirical Formula (Hill Notation):
C6H13N3O2S
CAS Number:
Molecular Weight:
191.25

Products

Brand
Formula Weight
Gene Alias
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing