
Tetrakis(diethylamido)titanium(IV)

Synonym(s):
TDEAT, Tetrakis(diethylamino)titanium(IV), Titanium(IV) tetrakis(diethylamide)
Linear Formula:
[(C2H5)2N]4Ti
CAS Number:
Molecular Weight:
336.38
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing