Skip to Content
Merck
tert-Butyl trimethylsilylacetate ≥95.0% (GC)

tert-Butyl trimethylsilylacetate

tert-Butyl trimethylsilylacetate ≥95.0% (GC)
Empirical Formula (Hill Notation):
C9H20O2Si
CAS Number:
Molecular Weight:
188.34
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing