![tert-Butyl ethyl malonate 95%](/deepweb/assets/sigmaaldrich/product/structures/403/066/ffcb3e7e-0bab-49e1-ad5c-640d9f5dc98d/640/ffcb3e7e-0bab-49e1-ad5c-640d9f5dc98d.png)
tert-Butyl ethyl malonate
![tert-Butyl ethyl malonate 95%](/deepweb/assets/sigmaaldrich/product/structures/403/066/ffcb3e7e-0bab-49e1-ad5c-640d9f5dc98d/640/ffcb3e7e-0bab-49e1-ad5c-640d9f5dc98d.png)
Linear Formula:
(CH3)3CO2CCH2CO2C2H5
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing