Skip to Content
Merck
tert-Butyldimethylsilyl cyanide 97%

tert-Butyldimethylsilyl cyanide

tert-Butyldimethylsilyl cyanide 97%
Synonym(s):
tert-Butyl-cyano-dimethylsilane
Linear Formula:
(CH3)3CSi(CH3)2CN
CAS Number:
Molecular Weight:
141.29

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing