tert-Butyl bicyclo[2.2.2]phosphorothionate
Synonym(s):
TBPS
Empirical Formula (Hill Notation):
C8H15O3PS
CAS Number:
Molecular Weight:
222.24
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing