Skip to Content
Merck
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate 95%

tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate

tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate 95%
Empirical Formula (Hill Notation):
C10H17F2NO4
CAS Number:
Molecular Weight:
253.24

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing