Skip to Content
Merck
Sulprofos PESTANAL®, analytical standard

Sulprofos

Sulprofos PESTANAL®, analytical standard
Synonym(s):
Sulprophos
Empirical Formula (Hill Notation):
C12H19O2PS3
CAS Number:
Molecular Weight:
322.45
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing