![Sulforhodamine 101 acid chloride Technical grade](/deepweb/assets/sigmaaldrich/product/structures/640/209/16b4f405-6aad-40ee-ae26-b04e2af76467/640/16b4f405-6aad-40ee-ae26-b04e2af76467.png)
Sulforhodamine 101 acid chloride
![Sulforhodamine 101 acid chloride Technical grade](/deepweb/assets/sigmaaldrich/product/structures/640/209/16b4f405-6aad-40ee-ae26-b04e2af76467/640/16b4f405-6aad-40ee-ae26-b04e2af76467.png)
Synonym(s):
Sulforhodamine 101 sulfonyl chloride
Empirical Formula (Hill Notation):
C31H29ClN2O6S2
CAS Number:
Molecular Weight:
625.15
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing