Skip to Content
Merck
Spiro[2.4]hepta-4,6-diene ≥95.0% (GC)

Spiro[2.4]hepta-4,6-diene

Spiro[2.4]hepta-4,6-diene ≥95.0% (GC)
Empirical Formula (Hill Notation):
C7H8
CAS Number:
Molecular Weight:
92.14

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing