Sodium mesoxalate monohydrate
Synonym(s):
Dihydroxymalonic acid disodium salt, Ketomalonic acid disodium salt, Mesoxalic acid disodium salt monohydrate
Linear Formula:
(HO)2C(COONa)2
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing