Skip to Content
VN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver nanoplates
Products
Brand
Sigma-Aldrich (6)
Physical Form
Functional Group
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
807680
Silver nanoplates,
resonant wavelength, 950 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Expand
View Pricing
807516
Silver nanoplates,
resonant wavelength, 750 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Expand
View Pricing
807532
Silver nanoplates,
resonant wavelength, 550 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Expand
View Pricing
807699
Silver nanoplates,
resonant wavelength, 850 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Expand
View Pricing
807672
Silver nanoplates,
resonant wavelength, 1050 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Expand
View Pricing
807524
Silver nanoplates,
resonant wavelength, 650 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Expand
View Pricing