Skip to Content
Merck
(S)-(−)-Glycidol 97%, optical purity ee: 98% (GLC)

(S)-(−)-Glycidol

(S)-(−)-Glycidol 97%, optical purity ee: 98% (GLC)
Synonym(s):
(S)-(−)-2,3-Epoxy-1-propanol, (S)-(−)-Oxirane-2-methanol
Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing