Skip to Content
Merck
(S)-(+)-4-Phenyl-2-oxazolidinone 98%

(S)-(+)-4-Phenyl-2-oxazolidinone

(S)-(+)-4-Phenyl-2-oxazolidinone 98%
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing