Skip to Content
Merck
(S)-(+)-3,5-Dinitro-N-(1-phenylethyl)benzamide 98%

(S)-(+)-3,5-Dinitro-N-(1-phenylethyl)benzamide

(S)-(+)-3,5-Dinitro-N-(1-phenylethyl)benzamide 98%
Synonym(s):
(S)-(+)-N-(3,5-Dinitrobenzoyl)-α-methylbenzylamine, (S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
Linear Formula:
C6H5CH(CH3)NHCOC6H3(NO2)2
CAS Number:
Molecular Weight:
315.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing