(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol
Synonym(s):
(S)-(+)-α-Tetralol
Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing