Skip to Content
Merck
(S)-(−)-α-Methyl-4-pyridinemethanol 99%

(S)-(−)-α-Methyl-4-pyridinemethanol

(S)-(−)-α-Methyl-4-pyridinemethanol 99%
Synonym(s):
(S)-(−)-1-(4-Pyridyl)ethanol
Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing