Skip to Content
Merck
Samarium(III) trifluoromethanesulfonate 98%

Samarium(III) trifluoromethanesulfonate

Samarium(III) trifluoromethanesulfonate 98%
Synonym(s):
Samarium(III) triflate, Trifluoromethanesulfonic acid samarium(III) salt
Linear Formula:
Sm(SO3CF3)3
CAS Number:
Molecular Weight:
597.57

Products

Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing