
Samarium(III) sulfate octahydrate

Linear Formula:
Sm2(SO4)3 · 8H2O
CAS Number:
Molecular Weight:
733.03
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing