Skip to Content
Merck
(S)-(−)-4-Isopropyl-2-oxazolidinethione 99%

(S)-(−)-4-Isopropyl-2-oxazolidinethione

(S)-(−)-4-Isopropyl-2-oxazolidinethione 99%
Empirical Formula (Hill Notation):
C6H11NOS
CAS Number:
Molecular Weight:
145.22

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing