
(S)-2-(Boc-amino)-3-phenyl-1-propanol

Synonym(s):
(S)-(−)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, N-(tert-Butoxycarbonyl)-L-phenylalaninol, Boc-L-phenylalaninol
Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
251.32
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing