Skip to Content
Merck
(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthalene 97%

(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthalene

(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthalene 97%
Synonym(s):
(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthyl, (S)-1,1′-Bi-2-naphthyl dimethyl ether
Empirical Formula (Hill Notation):
C22H18O2
CAS Number:
Molecular Weight:
314.38

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing