![(S)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine 97%](/deepweb/assets/sigmaaldrich/product/structures/831/826/11a39975-c391-41f5-a2c2-a90619f503f1/640/11a39975-c391-41f5-a2c2-a90619f503f1.png)
(S)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine
![(S)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine 97%](/deepweb/assets/sigmaaldrich/product/structures/831/826/11a39975-c391-41f5-a2c2-a90619f503f1/640/11a39975-c391-41f5-a2c2-a90619f503f1.png)
Synonym(s):
(S)-1-Methyl-2-(piperidinomethyl)pyrrolidine
Empirical Formula (Hill Notation):
C11H22N2
CAS Number:
Molecular Weight:
182.31
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing