
(R,R)-(+)-Hydrobenzoin

Synonym(s):
(R,R)-1,2-Diphenyl-1,2-ethanediol, (R,R)-1,2-Diphenylethylene glycol
Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing