Skip to Content
Merck
(R)-(−)-Hexahydromandelic acid 98%, optical purity ee: 99% (GLC)

(R)-(−)-Hexahydromandelic acid

(R)-(−)-Hexahydromandelic acid 98%, optical purity ee: 99% (GLC)
Synonym(s):
(R)-(−)-Cyclohexylhydroxyacetic acid
Linear Formula:
C6H11CH(OH)CO2H
CAS Number:
Molecular Weight:
158.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing