Skip to Content
Merck
Recombinant CRM197 expressed in Pseudomonas fluorescens, [Glu52]- Diphtheria toxin lyophilized powder

Recombinant CRM197 expressed in Pseudomonas fluorescens, [Glu52]- Diphtheria toxin

Recombinant CRM197 expressed in Pseudomonas fluorescens, [Glu52]- Diphtheria toxin lyophilized powder
Synonym(s):
[Glu52]-Diphtheria toxin from Corynebacterium diphtheriae, CRM 197
CAS Number:

Products

Product Category
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing