Skip to Content
Merck
(R)-(+)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-2-naphthol 95%

(R)-(+)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-2-naphthol

(R)-(+)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-2-naphthol 95%
Linear Formula:
(C10H10OH)2
CAS Number:
Molecular Weight:
294.39

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing