Skip to Content
Merck
(R)-(+)-1-(4-Bromophenyl)ethylamine ≥96.0% (sum of enantiomers, GC)

(R)-(+)-1-(4-Bromophenyl)ethylamine

(R)-(+)-1-(4-Bromophenyl)ethylamine ≥96.0% (sum of enantiomers, GC)
Synonym(s):
(R)-(+)-4-Bromo-α-methylbenzylamine
Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing