Skip to Content
VN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rare earth doped phosphor nanoparticles
Products
Brand
Sigma-Aldrich (4)
Physical Form
Chemical Composition
Functional Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
900558
Rare earth doped phosphor nanoparticles,
fluorescence λ
em
545 nm (main peak), 50 mg/mL in H
2
O
Expand
View Pricing
900557
Rare earth doped phosphor nanoparticles,
fluorescence λ
em
620 nm (main peak), 50 mg/mL in H
2
O
Expand
View Pricing
900559
Rare earth doped phosphor nanoparticles,
fluorescence λ
em
390 nm (main peak), 50 mg/mL in H
2
O
Expand
View Pricing
900591
Rare earth doped phosphor nanoparticles,
fluorescence λ
em
580 nm (main peak), 50 mg/mL in H
2
O
Expand
View Pricing