Skip to Content
Merck
(R)-(4-Fluorophenyl)oxirane technical, ≥90.0% (GC)

(R)-(4-Fluorophenyl)oxirane

(R)-(4-Fluorophenyl)oxirane technical, ≥90.0% (GC)
Synonym(s):
(R)-4-Fluorostyrene oxide
Empirical Formula (Hill Notation):
C8H7FO
CAS Number:
Molecular Weight:
138.14

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing