Skip to Content
Merck
(R)-(−)-3-Aminopiperidine dihydrochloride ≥98.0% (GC)

(R)-(−)-3-Aminopiperidine dihydrochloride

(R)-(−)-3-Aminopiperidine dihydrochloride ≥98.0% (GC)
Synonym(s):
(R)-3-Piperidinamine dihydrochloride
Empirical Formula (Hill Notation):
C5H12N2 · 2HCl
CAS Number:
Molecular Weight:
173.08

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing