Skip to Content
Merck
(R)-2-Hydroxy-4-phenylbutyric acid 99% (sum of enantiomers)

(R)-2-Hydroxy-4-phenylbutyric acid

(R)-2-Hydroxy-4-phenylbutyric acid 99% (sum of enantiomers)
Linear Formula:
C6H5CH2CH2CH(OH)CO2H
CAS Number:
Molecular Weight:
180.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing