Skip to Content
Merck
(R)-(−)-1,2,3,4-Tetrahydro-1-naphthol 99%

(R)-(−)-1,2,3,4-Tetrahydro-1-naphthol

(R)-(−)-1,2,3,4-Tetrahydro-1-naphthol 99%
Synonym(s):
(R)-(−)-α-Tetralol
Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing