![p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone 98%](/deepweb/assets/sigmaaldrich/product/structures/220/490/3786cdd8-0453-4290-9a28-5060d0657071/640/3786cdd8-0453-4290-9a28-5060d0657071.png)
p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone
![p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone 98%](/deepweb/assets/sigmaaldrich/product/structures/220/490/3786cdd8-0453-4290-9a28-5060d0657071/640/3786cdd8-0453-4290-9a28-5060d0657071.png)
Synonym(s):
1-(p-Toluenesulfonyl)-2-(trimethylsilyl)acetylene, 2-(Trimethylsilyl)ethynyl p-tolyl sulfone
Linear Formula:
(CH3)3SiC≡CSO2C6H4CH3
CAS Number:
Molecular Weight:
252.40
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing