Profoxydim lithium salt
Synonym(s):
Clefoxydim lithium salt
Empirical Formula (Hill Notation):
C24H31ClLiNO4S
CAS Number:
Molecular Weight:
471.97
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing