Skip to Content
Merck
Potassium hexafluoronickelate(IV) 99%

Potassium hexafluoronickelate(IV)

Potassium hexafluoronickelate(IV) 99%
Synonym(s):
Potassium nickel(IV) fluoride
Linear Formula:
K2NiF6
CAS Number:
Molecular Weight:
250.88

Products

Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing