
Potassium 3-methoxyphenyltrifluoroborate

Linear Formula:
H3COC6H4BF3 ·K
CAS Number:
Molecular Weight:
214.03
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing