![Poly[(9,9-di-(2-ethylhexyl)-9H-fluorene-2,7-vinylene)-co-(1-methoxy-4-(2-ethylhexyloxy)-2,5-phenylenevinylene)] (90:10 mole ratio)](/deepweb/assets/sigmaaldrich/product/structures/754/750/097b4f13-c2b2-4d47-bb9f-4e238448bf5b/640/097b4f13-c2b2-4d47-bb9f-4e238448bf5b.png)
Poly[(9,9-di-(2-ethylhexyl)-9H-fluorene-2,7-vinylene)-co-(1-methoxy-4-(2-ethylhexyloxy)-2,5-phenylenevinylene)]
![Poly[(9,9-di-(2-ethylhexyl)-9H-fluorene-2,7-vinylene)-co-(1-methoxy-4-(2-ethylhexyloxy)-2,5-phenylenevinylene)] (90:10 mole ratio)](/deepweb/assets/sigmaaldrich/product/structures/754/750/097b4f13-c2b2-4d47-bb9f-4e238448bf5b/640/097b4f13-c2b2-4d47-bb9f-4e238448bf5b.png)
Synonym(s):
PFV
Linear Formula:
(C31H42)m(C17H24O2)n
Products
Brand
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing