Skip to Content
Merck
p-Iodo-D-phenylalanine 97%

p-Iodo-D-phenylalanine

p-Iodo-D-phenylalanine 97%
Synonym(s):
2-Amino-3-(4-iodophenyl)-propanoic acid
Linear Formula:
C9H10NO2I
CAS Number:
Molecular Weight:
291.09

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing