
Phorbol 12-myristate 13-acetate 4-O-methyl ether

Synonym(s):
MPMA
Empirical Formula (Hill Notation):
C37H58O8
CAS Number:
Molecular Weight:
630.85
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing