
Phenoxymethylpenicillinic acid potassium salt

Synonym(s):
Penicillin V potassium salt
Empirical Formula (Hill Notation):
C16H17N2O5SK
Molecular Weight:
388.48
EC Number:
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing