Skip to Content
Merck
Perfluorotetracosane analytical standard, suitable for mass spectrometry (MS)

Perfluorotetracosane

Perfluorotetracosane analytical standard, suitable for mass spectrometry (MS)
Linear Formula:
CF3(CF2)22CF3
CAS Number:
Molecular Weight:
1238.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing