Skip to Content
Merck
Oroxin B ≥98% (HPLC)

Oroxin B

Oroxin B ≥98% (HPLC)
Synonym(s):
5,6,7-Trihydroxyflavone 7-diglucoside, Baicalein 7-gentiobioside, Baicalein-7-O-diglucoside
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing