Skip to Content
Merck
Oleic anhydride ≥95.0% (GC)

Oleic anhydride

Oleic anhydride ≥95.0% (GC)
Synonym(s):
cis-9-Octadecenoic anhydride
Linear Formula:
[CH3(CH2)7CH=CH(CH2)7CO]2O
CAS Number:
Molecular Weight:
546.91
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing