O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N′,N′-tetramethyluronium tetrafluoroborate
Synonym(s):
N-[[[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy](dimethylamino)methylene]-N-methyl-methanaminium tetrafluoroborate, TOTU
Linear Formula:
C2H5O2CC(CN)=NOC[N(CH3)2]=N(CH3)2BF4
CAS Number:
Molecular Weight:
328.07
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing