Skip to Content
Merck
O-Allyl 2,2,2-trichloroacetimidate 96%

O-Allyl 2,2,2-trichloroacetimidate

O-Allyl 2,2,2-trichloroacetimidate 96%
Synonym(s):
2,2,2-Trichloroacetimidic acid allyl ester
Empirical Formula (Hill Notation):
C5H6Cl3NO
CAS Number:
Molecular Weight:
202.47

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing