Skip to Content
Merck
O-(4-Chlorobenzoyl)hydroquinine 98%

O-(4-Chlorobenzoyl)hydroquinine

O-(4-Chlorobenzoyl)hydroquinine 98%
Synonym(s):
Dihydroquinine 4-chlorobenzoate, Hydroquinine 4-chlorobenzoate
Empirical Formula (Hill Notation):
C27H29ClN2O3
CAS Number:
Molecular Weight:
464.98

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing