Skip to Content
Merck
N-(tert-Butoxycarbonyl)-L-proline N′-methoxy-N′-methylamide 98%

N-(tert-Butoxycarbonyl)-L-proline N′-methoxy-N′-methylamide

N-(tert-Butoxycarbonyl)-L-proline N′-methoxy-N′-methylamide 98%
Empirical Formula (Hill Notation):
C12H22N2O4
CAS Number:
Molecular Weight:
258.31

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing